The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015136.2 | NP_055951.2 | 9 - 10 | 1097 | 86 |
XM_005264973.3 | XP_005265030.1 | 9 - 10 | 1099 | 86 | |
XM_005264974.2 | XP_005265031.1 | 9 - 10 | 1099 | 86 | |
XM_006713065.1 | XP_006713128.1 | 9 - 10 | 1099 | 86 | |
XM_017005998.1 | XP_016861487.1 | 9 - 10 | 1099 | 86 | |
XM_017005999.1 | XP_016861488.1 | 9 - 10 | 1099 | 86 | |
XM_017006000.1 | XP_016861489.1 | 9 - 10 | 1099 | 86 | |
XM_017006001.1 | XP_016861490.1 | 9 - 10 | 1099 | 86 | |
XM_017006002.1 | XP_016861491.1 | 9 - 10 | 1099 | 86 | |
XM_017006003.1 | XP_016861492.1 | 9 - 10 | 1099 | 86 | |
XM_017006004.1 | XP_016861493.1 | 9 - 10 | 1099 | 86 | |
GenBank mRNA | AB052956.1 | - | 9 - 10 | 1084 | 86 |
AB052957.1 | - | 9 - 10 | 1078 | 86 | |
AJ275213.1 | - | 9 - 10 | 1053 | 86 | |
DQ786774.1 | - | 9 - 10 | 1047 | 86 |