Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GRAMD1B
- Entrez Gene ID
-
57476
- Gene Name
- GRAM domain containing 1B
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 123358299 - 123627771 on Build GRCh38
View Full Product Details