Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRAMD1B
Entrez Gene ID
57476
Gene Name
GRAM domain containing 1B
Gene Aliases
-
Chromosome Location
Chr.11: 123358299 - 123627771 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286563.1 NP_001273492.1 7 - 8 1074 59
NM_001286564.1 NP_001273493.1 6 - 7 783 59
NM_020716.2 NP_065767.1 6 - 7 1053 59
XM_005271619.4 XP_005271676.1 6 - 7 1053 59
XM_005271621.4 XP_005271678.1 7 - 8 5440 59
XM_005271623.4 XP_005271680.1 6 - 7 818 59
XM_005271624.4 XP_005271681.2 6 - 7 733 59
XM_006718892.3 XP_006718955.1 7 - 8 970 59
XM_011542926.2 XP_011541228.1 7 - 8 967 59
XM_011542929.2 XP_011541231.1 6 - 7 943 59
XM_011542930.2 XP_011541232.1 6 - 7 1584 59
XM_011542931.2 XP_011541233.1 6 - 7 1498 59
XM_011542933.1 XP_011541235.1 4 - 5 1195 59
XM_017018034.1 XP_016873523.1 8 - 9 1038 59
XM_017018035.1 XP_016873524.1 8 - 9 1035 59
XM_017018036.1 XP_016873525.1 7 - 8 1505 59
XM_017018037.1 XP_016873526.1 8 - 9 1804 59
XM_017018038.1 XP_016873527.1 8 - 9 1038 59
XM_017018039.1 XP_016873528.1 7 - 8 1096 59
XM_017018040.1 XP_016873529.1 6 - 7 892 59
XM_017018041.1 XP_016873530.1 7 - 8 1096 59
XM_017018042.1 XP_016873531.1 7 - 8 1096 59
XM_017018043.1 XP_016873532.1 6 - 7 729 59
XM_017018044.1 XP_016873533.1 6 - 7 1053 59
XM_017018045.1 XP_016873534.1 7 - 8 5437 59
XM_017018046.1 XP_016873535.1 7 - 8 1542 59
XM_017018047.1 XP_016873536.1 7 - 8 1539 59
XM_017018048.1 XP_016873537.1 6 - 7 1495 59
XM_017018049.1 XP_016873538.1 7 - 8 1536 59
XM_017018050.1 XP_016873539.1 6 - 7 812 59
XM_017018051.1 XP_016873540.1 4 - 5 416 59
GenBank mRNA AB033027.1 - 6 - 7 716 59
AK127457.1 - 7 - 8 1074 59
AK297606.1 - 6 - 7 783 59
AK307795.1 - 6 - 7 893 59
BC105741.1 - 6 - 7 867 59
BC105931.1 - 6 - 7 867 59
BC105932.1 - 6 - 7 867 59
BC107480.1 - 6 - 7 867 59

View Full Product Details