The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173653.3 | NP_775924.1 | 3 - 4 | 663 | 112 |
XM_011512704.2 | XP_011511006.1 | 3 - 4 | 636 | 112 | |
XM_017006202.1 | XP_016861691.1 | 3 - 4 | 636 | 112 | |
XM_017006203.1 | XP_016861692.1 | 2 - 3 | 421 | 112 | |
GenBank mRNA | AB089794.1 | - | 3 - 4 | 454 | 112 |
AK301816.1 | - | 2 - 3 | 389 | 112 | |
AK315107.1 | - | 3 - 4 | 590 | 112 | |
AL832304.1 | - | 3 - 4 | 528 | 112 | |
AY254100.1 | - | 3 - 4 | 578 | 112 | |
BC035779.1 | - | 3 - 4 | 579 | 112 |