Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC9A9
Entrez Gene ID
285195
Gene Name
solute carrier family 9 member A9
Gene Aliases
AUTS16, NHE9
Chromosome Location
Chr.3: 143265222 - 143848531 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173653.3 NP_775924.1 4 - 5 738 79
XM_011512704.2 XP_011511006.1 4 - 5 711 79
XM_017006202.1 XP_016861691.1 4 - 5 711 79
XM_017006203.1 XP_016861692.1 3 - 4 496 79
GenBank mRNA AB089794.1 - 4 - 5 529 79
AK301816.1 - 3 - 4 464 79
AK315107.1 - 4 - 5 665 79
AL832304.1 - 4 - 5 603 79
AY254100.1 - 4 - 5 653 79
BC035779.1 - 4 - 5 654 79

View Full Product Details