Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPXM2
Entrez Gene ID
119587
Gene Name
carboxypeptidase X (M14 family), member 2
Gene Aliases
CPX2, UNQ676
Chromosome Location
Chr.10: 123745636 - 123944094 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198148.2 NP_937791.2 8 - 9 1428 75
XM_005269528.3 XP_005269585.1 8 - 9 1278 75
XM_011539283.2 XP_011537585.1 8 - 9 1278 75
XM_011539285.2 XP_011537587.1 8 - 9 1060 75
XM_011539286.1 XP_011537588.1 7 - 8 1002 75
XM_017015673.1 XP_016871162.1 9 - 10 1029 75
XM_017015674.1 XP_016871163.1 2 - 3 204 75
GenBank mRNA AK296365.1 - 8 - 9 835 75
AK300506.1 - 4 - 5 389 75
AK304023.1 - 8 - 9 1202 75
AK304814.1 - 8 - 9 1239 75
AL833732.1 - 6 - 7 606 75
AY358565.1 - 8 - 9 1428 75
BC036789.1 - 8 - 9 1258 75
BC137492.1 - 8 - 9 1423 75
BC146862.1 - 8 - 9 1423 75

View Full Product Details