The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007115.3 | NP_009046.2 | 2 - 3 | 312 | 77 |
GenBank mRNA | AF086484.1 | - | 2 - 3 | 164 | 77 |
AJ419936.1 | - | 2 - 3 | 237 | 77 | |
AJ421518.1 | - | 2 - 3 | 237 | 77 | |
BC030205.1 | - | 2 - 3 | 281 | 77 | |
M31165.1 | - | 2 - 3 | 305 | 77 |