Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NINL
Entrez Gene ID
22981
Gene Name
ninein like
Gene Aliases
NLP
Chromosome Location
Chr.20: 25452697 - 25585531 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318226.1 NP_001305155.1 14 - 15 1899 133
NM_025176.5 NP_079452.3 14 - 15 1899 133
XM_011529186.2 XP_011527488.1 14 - 15 2537 133
XM_011529187.2 XP_011527489.1 14 - 15 2522 133
XM_011529188.1 XP_011527490.1 14 - 15 1914 133
XM_011529189.1 XP_011527491.1 14 - 15 1922 133
XM_011529190.1 XP_011527492.1 13 - 14 1837 133
XM_011529191.1 XP_011527493.1 10 - 11 1429 133
XM_011529192.1 XP_011527494.1 7 - 8 2349 133
XM_011529193.1 XP_011527495.1 13 - 14 1837 133
XM_011529194.2 XP_011527496.1 5 - 6 1618 133
XM_011529195.2 XP_011527497.1 4 - 5 443 133
XM_011529197.2 XP_011527499.1 6 - 7 812 133
XM_017027732.1 XP_016883221.1 15 - 16 3856 133
XM_017027733.1 XP_016883222.1 15 - 16 1968 133
XM_017027734.1 XP_016883223.1 8 - 9 1003 133
XM_017027735.1 XP_016883224.1 4 - 5 448 133
GenBank mRNA AB023197.1 - 14 - 15 1885 133
AK093569.1 - 4 - 5 1282 133
AK309298.1 - 14 - 15 1848 133
BC036380.2 - 13 - 14 1837 133
EU718622.1 - 14 - 15 1885 133

View Full Product Details