The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001082533.1 | NP_001076002.1 | 2 - 3 | 1174 | 67 |
NM_001082534.1 | NP_001076003.1 | 2 - 3 | 739 | 67 | |
NM_020178.4 | NP_064563.1 | 1 - 2 | 1048 | 67 | |
GenBank mRNA | AB001597.1 | - | 1 - 2 | 62 | 67 |
AB036836.1 | - | 2 - 3 | 964 | 67 | |
AF288385.1 | - | 2 - 3 | 730 | 67 | |
AK313001.1 | - | 1 - 2 | 624 | 67 | |
AL161812.1 | - | 1 - 2 | 64 | 67 | |
AY358509.1 | - | 1 - 2 | 342 | 67 | |
BC020577.1 | - | 2 - 3 | 706 | 67 | |
BC029865.1 | - | 2 - 3 | 708 | 67 | |
BC047456.1 | - | 1 - 2 | 961 | 67 | |
BC068462.1 | - | 1 - 2 | 1007 | 67 |