Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRSS42
Entrez Gene ID
339906
Gene Name
protease, serine 42
Gene Aliases
TESSP2
Chromosome Location
Chr.3: 46830404 - 46834095 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK092703.1 - 2 - 3 331 85

View Full Product Details