Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC28
Entrez Gene ID
23331
Gene Name
tetratricopeptide repeat domain 28
Gene Aliases
TPRBK
Chromosome Location
Chr.22: 27978014 - 28679865 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145418.1 NP_001138890.1 15 - 16 4540 76
XM_005261405.2 XP_005261462.1 15 - 16 4540 76
XM_006724171.3 XP_006724234.1 14 - 15 13318 76
XM_011530018.2 XP_011528320.1 15 - 16 4719 76
XM_017028673.1 XP_016884162.1 15 - 16 4513 76
GenBank mRNA AB028966.3 - 10 - 11 3333 76
AB665284.1 - 15 - 16 4488 76
BC016465.1 - 7 - 8 1084 76

View Full Product Details