The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006620.1 | NP_001006621.1 | 1 - 2 | 264 | 60 |
GenBank mRNA | AL833042.1 | - | 1 - 2 | 225 | 60 |
M37191.1 | - | 1 - 2 | 37 | 60 |