The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006617.1 | NP_001006618.1 | 9 - 10 | 1535 | 84 |
NM_001006620.1 | NP_001006621.1 | 8 - 9 | 1207 | 84 | |
NM_001006621.1 | NP_001006622.1 | 7 - 8 | 1117 | 84 | |
NM_024117.3 | NP_077022.1 | 8 - 9 | 1427 | 84 | |
XM_011519005.1 | XP_011517307.1 | 10 - 11 | 1676 | 84 | |
XM_011519006.2 | XP_011517308.1 | 9 - 10 | 1568 | 84 | |
XM_011519010.2 | XP_011517312.1 | 6 - 7 | 853 | 84 | |
XM_017015126.1 | XP_016870615.1 | 9 - 10 | 1424 | 84 | |
XM_017015127.1 | XP_016870616.1 | 8 - 9 | 1316 | 84 | |
GenBank mRNA | AK290060.1 | - | 8 - 9 | 1240 | 84 |
AK295364.1 | - | 6 - 7 | 841 | 84 | |
AK298841.1 | - | 4 - 5 | 501 | 84 | |
AK310361.1 | - | 5 - 6 | 733 | 84 | |
AK316149.1 | - | 6 - 7 | 780 | 84 | |
AL833042.1 | - | 8 - 9 | 1168 | 84 | |
AY524429.1 | - | 9 - 10 | 1532 | 84 | |
AY524431.1 | - | 8 - 9 | 1424 | 84 | |
AY633624.1 | - | 8 - 9 | 1394 | 84 | |
AY633625.1 | - | 9 - 10 | 1509 | 84 | |
AY633626.1 | - | 10 - 11 | 1632 | 84 | |
BC002326.2 | - | 8 - 9 | 1396 | 84 | |
BC003044.1 | - | 8 - 9 | 1396 | 84 | |
M37191.1 | - | 8 - 9 | 980 | 84 |