Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC44A3
Entrez Gene ID
126969
Gene Name
solute carrier family 44 member 3
Gene Aliases
CTL3
Chromosome Location
Chr.1: 94820342 - 94895247 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114106.2 NP_001107578.1 6 - 7 779 79
NM_001258340.1 NP_001245269.1 6 - 7 779 79
NM_001258341.1 NP_001245270.1 6 - 7 683 79
NM_001258342.1 NP_001245271.1 5 - 6 671 79
NM_001258343.1 NP_001245272.1 5 - 6 575 79
NM_001301079.1 NP_001288008.1 5 - 6 618 79
NM_152369.4 NP_689582.2 5 - 6 714 79
XM_005270440.2 XP_005270497.1 6 - 7 674 79
XM_005270441.2 XP_005270498.1 6 - 7 716 79
XM_005270442.1 XP_005270499.1 6 - 7 770 79
XM_011540632.2 XP_011538934.1 5 - 6 643 79
XM_011540633.2 XP_011538935.1 5 - 6 481 79
XM_011540635.2 XP_011538937.1 6 - 7 770 79
XM_011540636.2 XP_011538938.1 6 - 7 674 79
XM_017000262.1 XP_016855751.1 5 - 6 656 79
XM_017000263.1 XP_016855752.1 6 - 7 716 79
XM_017000264.1 XP_016855753.1 6 - 7 770 79
XM_017000265.1 XP_016855754.1 6 - 7 674 79
XM_017000266.1 XP_016855755.1 6 - 7 770 79
GenBank mRNA AK300193.1 - 5 - 6 512 79
AK301288.1 - 5 - 6 520 79
AK302381.1 - 3 - 4 363 79
AK303906.1 - 5 - 6 650 79
AK313574.1 - 6 - 7 667 79
AK316123.1 - 7 - 8 624 79
AL530322.3 - 6 - 7 771 79
AY358659.1 - 2 - 3 233 79
BC033858.1 - 6 - 7 742 79
BC053877.1 - 5 - 6 637 79

View Full Product Details