Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC44A3
Entrez Gene ID
126969
Gene Name
solute carrier family 44 member 3
Gene Aliases
CTL3
Chromosome Location
Chr.1: 94820342 - 94895247 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114106.2 NP_001107578.1 8 - 9 993 150
NM_001258340.1 NP_001245269.1 8 - 9 993 150
NM_001258341.1 NP_001245270.1 8 - 9 897 150
NM_001258342.1 NP_001245271.1 7 - 8 885 150
NM_001258343.1 NP_001245272.1 7 - 8 789 150
NM_001301079.1 NP_001288008.1 7 - 8 832 150
NM_152369.4 NP_689582.2 7 - 8 928 150
XM_005270440.2 XP_005270497.1 8 - 9 888 150
XM_005270441.2 XP_005270498.1 8 - 9 930 150
XM_005270442.1 XP_005270499.1 8 - 9 984 150
XM_011540632.2 XP_011538934.1 7 - 8 857 150
XM_011540633.2 XP_011538935.1 7 - 8 695 150
XM_011540635.2 XP_011538937.1 8 - 9 984 150
XM_011540636.2 XP_011538938.1 8 - 9 888 150
XM_017000262.1 XP_016855751.1 7 - 8 870 150
XM_017000263.1 XP_016855752.1 8 - 9 930 150
XM_017000264.1 XP_016855753.1 8 - 9 984 150
XM_017000265.1 XP_016855754.1 8 - 9 888 150
XM_017000266.1 XP_016855755.1 8 - 9 984 150
GenBank mRNA AK300193.1 - 7 - 8 726 150
AK302381.1 - 5 - 6 577 150
AK303906.1 - 7 - 8 864 150
AK313574.1 - 8 - 9 881 150
AK316123.1 - 9 - 10 838 150
AY358659.1 - 4 - 5 447 150
BC033858.1 - 8 - 9 956 150
BC053877.1 - 7 - 8 851 150
BX648225.1 - 7 - 8 857 150

View Full Product Details