The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134758.3 | NP_001128230.1 | 1 - 2 | 153 | 89 |
NM_001267545.2 | NP_001254474.1 | 1 - 2 | 153 | 89 | |
NM_003224.5 | NP_003215.1 | 1 - 2 | 153 | 89 | |
NR_051954.2 | 1 - 2 | 153 | 89 | ||
NR_051957.2 | 1 - 2 | 153 | 89 | ||
NR_051958.2 | 1 - 2 | 153 | 89 | ||
GenBank mRNA | AL533893.3 | - | 1 - 2 | 143 | 89 |
BC021023.1 | - | 1 - 2 | 38 | 89 | |
BC093967.1 | - | 1 - 2 | 56 | 89 | |
BC143447.1 | - | 1 - 2 | 56 | 89 | |
BI524032.1 | - | 1 - 2 | 152 | 89 | |
BM549361.1 | - | - | 121 | 89 | |
BX342690.2 | - | 1 - 2 | 39 | 89 | |
BX342805.2 | - | 1 - 2 | 132 | 89 |