Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO3
Entrez Gene ID
63982
Gene Name
anoctamin 3
Gene Aliases
C11orf25, DYT23, DYT24, GENX-3947, TMEM16C
Chromosome Location
Chr.11: 26189123 - 26663289 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313726.1 NP_001300655.1 21 - 22 2273 83
NM_001313727.1 NP_001300656.1 17 - 18 1831 83
NM_031418.3 NP_113606.2 20 - 21 2181 83
XM_011520282.2 XP_011518584.1 7 - 8 728 83
XM_017018118.1 XP_016873607.1 18 - 19 1809 83
XM_017018119.1 XP_016873608.1 15 - 16 1421 83
GenBank mRNA AJ300461.1 - 20 - 21 2888 83
AK295143.1 - 17 - 18 1743 83
AK295658.1 - 4 - 5 216 83
AK295816.1 - 17 - 18 1743 83
AK302659.1 - 20 - 21 2220 83
AK304781.1 - 21 - 22 2114 83
BQ430769.1 - 1 - 2 52 83

View Full Product Details