Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO3
Entrez Gene ID
63982
Gene Name
anoctamin 3
Gene Aliases
C11orf25, DYT23, DYT24, GENX-3947, TMEM16C
Chromosome Location
Chr.11: 26189123 - 26663289 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313726.1 NP_001300655.1 23 - 24 2515 84
NM_001313727.1 NP_001300656.1 19 - 20 2073 84
NM_031418.3 NP_113606.2 22 - 23 2423 84
XM_011520282.2 XP_011518584.1 9 - 10 970 84
XM_017018118.1 XP_016873607.1 20 - 21 2051 84
XM_017018119.1 XP_016873608.1 17 - 18 1663 84
GenBank mRNA AJ300461.1 - 22 - 23 3130 84
AK295143.1 - 19 - 20 1985 84
AK295658.1 - 6 - 7 458 84
AK295816.1 - 19 - 20 1985 84
AK302659.1 - 22 - 23 2462 84
AK304781.1 - 23 - 24 2356 84
AK316018.1 - 19 - 20 1985 84
BQ430769.1 - 3 - 4 294 84

View Full Product Details