Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
A1BG
Entrez Gene ID
1
Gene Name
alpha-1-B glycoprotein
Gene Aliases
A1B, ABG, GAB, HYST2477
Chromosome Location
Chr.19: 58346806 - 58353499 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130786.3 NP_570602.2 3 - 4 405 81
GenBank mRNA AB073611.1 - 3 - 4 398 81
AK055885.1 - 3 - 4 398 81
AK289417.1 - 3 - 4 369 81
BC035719.1 - 3 - 4 452 81
BX325198.2 - 3 - 4 405 81
EU794585.1 - 3 - 4 397 81

View Full Product Details