The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042417.1 | NP_001035882.1 | 2 - 3 | 281 | 95 |
NM_017987.4 | NP_060457.4 | 2 - 3 | 608 | 95 | |
NR_103475.1 | 2 - 3 | 456 | 95 | ||
NR_103476.1 | 2 - 3 | 281 | 95 | ||
XM_005269953.4 | XP_005270010.1 | 2 - 3 | 608 | 95 | |
XM_005269955.4 | XP_005270012.1 | 1 - 2 | 187 | 95 | |
XM_005269956.4 | XP_005270013.1 | 2 - 3 | 608 | 95 | |
XM_005269957.4 | XP_005270014.1 | 2 - 3 | 278 | 95 | |
XM_006717911.3 | XP_006717974.1 | 2 - 3 | 279 | 95 | |
XM_011539942.2 | XP_011538244.1 | 2 - 3 | 1020 | 95 | |
GenBank mRNA | AB040970.1 | - | 2 - 3 | 217 | 95 |
AF411980.1 | - | 1 - 2 | 128 | 95 | |
AF461266.1 | - | 1 - 2 | 187 | 95 | |
AK127019.1 | - | 2 - 3 | 608 | 95 | |
AY528716.1 | - | 2 - 3 | 246 | 95 | |
BC041092.1 | - | 2 - 3 | 252 | 95 | |
DA858392.1 | - | 2 - 3 | 201 | 95 |