Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRD2
Entrez Gene ID
6046
Gene Name
bromodomain containing 2
Gene Aliases
D6S113E, FSH, FSRG1, NAT, O27.1.1, RING3, RNF3
Chromosome Location
Chr.6: 32968660 - 32981505 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113182.2 NP_001106653.1 3 - 4 1943 73
NM_001199455.1 NP_001186384.1 2 - 3 1633 73
NM_001199456.1 NP_001186385.1 2 - 3 346 73
NM_005104.3 NP_005095.1 3 - 4 2030 73
GenBank mRNA AK056504.1 - 3 - 4 1901 73
BC000477.2 - 2 - 3 1002 73
BC063840.1 - 2 - 3 498 73
BX647233.1 - 2 - 3 346 73
BX648109.1 - 3 - 4 1944 73
D42040.1 - 3 - 4 2030 73
M80613.1 - 2 - 3 1365 73
X62083.1 - 2 - 3 1365 73

View Full Product Details