Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRX1
Entrez Gene ID
79671
Gene Name
NLR family member X1
Gene Aliases
CLR11.3, DLNB26, NOD26, NOD5, NOD9
Chromosome Location
Chr.11: 119168334 - 119184016 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282143.1 NP_001269072.1 8 - 9 2586 79
NM_001282144.1 NP_001269073.1 8 - 9 2994 79
NM_001282358.1 NP_001269287.1 8 - 9 2518 79
NM_024618.3 NP_078894.2 8 - 9 2613 79
XM_005271669.1 XP_005271726.1 8 - 9 2837 79
XM_005271670.1 XP_005271727.1 7 - 8 2490 79
XM_005271671.1 XP_005271728.1 7 - 8 2664 79
XM_005271672.1 XP_005271729.1 7 - 8 2487 79
XM_006718904.1 XP_006718967.1 8 - 9 2871 79
XM_011542980.1 XP_011541282.1 8 - 9 2501 79
GenBank mRNA AB094095.1 - 8 - 9 2597 79
AK025131.1 - 3 - 4 1240 79
AK056454.1 - 7 - 8 2311 79
AK095247.1 - 6 - 7 1798 79
AK225223.1 - 3 - 4 1240 79
AK291716.1 - 8 - 9 2580 79
AY245437.1 - 7 - 8 2356 79
BC013199.1 - 4 - 5 1717 79
BC110890.1 - 8 - 9 2959 79
BX647705.1 - 8 - 9 2571 79
EF452237.1 - 7 - 8 2356 79

View Full Product Details