The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003129.3 | NP_003120.2 | 6 - 7 | 2036 | 147 |
XM_011517246.2 | XP_011515548.1 | 6 - 7 | 1134 | 147 | |
GenBank mRNA | AF098865.1 | - | 6 - 7 | 1265 | 147 |
AK225085.1 | - | 6 - 7 | 1326 | 147 | |
AK295302.1 | - | 6 - 7 | 936 | 147 | |
AK301792.1 | - | 6 - 7 | 928 | 147 | |
AK313384.1 | - | 6 - 7 | 1154 | 147 | |
BC017033.1 | - | 6 - 7 | 1268 | 147 | |
BX647605.1 | - | 6 - 7 | 2008 | 147 | |
D78129.1 | - | 4 - 5 | 554 | 147 | |
D78130.1 | - | 6 - 7 | 1373 | 147 |