The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_205849.3 | NP_995321.1 | 7 - 8 | 853 | 74 |
XM_011545464.2 | XP_011543766.1 | 14 - 15 | 1426 | 74 | |
GenBank mRNA | AF494344.1 | - | 7 - 8 | 852 | 74 |
AK097501.1 | - | 6 - 7 | 797 | 74 | |
AK128470.1 | - | 7 - 8 | 3214 | 74 | |
BC120955.1 | - | 6 - 7 | 552 | 74 | |
BC120956.1 | - | 6 - 7 | 552 | 74 |