Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRXN2
Entrez Gene ID
9379
Gene Name
neurexin 2
Gene Aliases
-
Chromosome Location
Chr.11: 64606174 - 64723188 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015080.3 NP_055895.1 11 - 12 2854 90
XM_005274400.3 XP_005274457.1 10 - 11 2386 90
XM_005274401.3 XP_005274458.1 10 - 11 2386 90
XM_005274402.3 XP_005274459.1 9 - 10 2362 90
XM_011545370.1 XP_011543672.1 10 - 11 2407 99
XM_011545371.1 XP_011543673.1 10 - 11 2404 99
XM_011545373.1 XP_011543675.1 10 - 11 2386 99
XM_011545375.1 XP_011543677.1 9 - 10 2362 99
XM_011545385.1 XP_011543687.1 3 - 4 752 99
XM_017018563.1 XP_016874052.1 9 - 10 2377 99
XM_017018564.1 XP_016874053.1 9 - 10 2374 99
XM_017018566.1 XP_016874055.1 9 - 10 2335 99
XM_017018567.1 XP_016874056.1 10 - 11 2407 99
XM_017018568.1 XP_016874057.1 10 - 11 2407 90
XM_017018569.1 XP_016874058.1 8 - 9 2305 99
XM_017018570.1 XP_016874059.1 10 - 11 2407 99
XM_017018571.1 XP_016874060.1 10 - 11 2407 90
XM_017018572.1 XP_016874061.1 10 - 11 1702 99
XM_017018574.1 XP_016874063.1 9 - 10 1672 99
XM_017018575.1 XP_016874064.1 9 - 10 1669 99
XM_017018576.1 XP_016874065.1 5 - 6 1317 99
XM_017018577.1 XP_016874066.1 3 - 4 752 90
XM_017018578.1 XP_016874067.1 10 - 11 2407 90
GenBank mRNA AB035266.1 - 10 - 11 2506 90
AB177862.1 - 7 - 8 1558 90

View Full Product Details