The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010875.3 | NP_001010875.1 | 5 - 6 | 606 | 134 |
NM_001286806.1 | NP_001273735.1 | 4 - 5 | 487 | 134 | |
NM_001286807.1 | NP_001273736.1 | 5 - 6 | 664 | 134 | |
XM_005266321.2 | XP_005266378.1 | 5 - 6 | 532 | 134 | |
XM_005266322.1 | XP_005266379.1 | 5 - 6 | 532 | 134 | |
XM_006719793.3 | XP_006719856.1 | 5 - 6 | 778 | 134 | |
XM_006719794.1 | XP_006719857.1 | 5 - 6 | 656 | 134 | |
XM_011535026.1 | XP_011533328.1 | 4 - 5 | 383 | 134 | |
XM_011535027.2 | XP_011533329.1 | 4 - 5 | 383 | 134 | |
XM_017020523.1 | XP_016876012.1 | 5 - 6 | 603 | 134 | |
XM_017020524.1 | XP_016876013.1 | 5 - 6 | 656 | 134 | |
GenBank mRNA | AK094133.1 | - | 5 - 6 | 533 | 134 |
AK095465.1 | - | 4 - 5 | 386 | 134 | |
AK302967.1 | - | 4 - 5 | 414 | 134 | |
BC132739.1 | - | 5 - 6 | 529 | 134 | |
BC136760.1 | - | 5 - 6 | 529 | 134 | |
BC144401.1 | - | 5 - 6 | 587 | 134 | |
DA184634.1 | - | 3 - 4 | 340 | 134 |