The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004784.2 | NP_004775.1 | 11 - 12 | 2692 | 81 |
XM_006714416.3 | XP_006714479.1 | 11 - 12 | 2748 | 81 | |
XM_017008839.1 | XP_016864328.1 | 11 - 12 | 2660 | 81 | |
XM_017008840.1 | XP_016864329.1 | 11 - 12 | 2530 | 81 | |
XM_017008841.1 | XP_016864330.1 | 10 - 11 | 4371 | 81 | |
XM_017008842.1 | XP_016864331.1 | 10 - 11 | 1975 | 81 | |
XM_017008843.1 | XP_016864332.1 | 8 - 9 | 1153 | 81 | |
XM_017008844.1 | XP_016864333.1 | 10 - 11 | 1410 | 81 | |
GenBank mRNA | AF074924.1 | - | 11 - 12 | 2692 | 81 |
AF076605.1 | - | 10 - 11 | 2478 | 81 | |
BC109309.1 | - | 11 - 12 | 2539 | 81 | |
BC109310.1 | - | 11 - 12 | 2539 | 81 |