The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004784.2 | NP_004775.1 | 5 - 6 | 1810 | 88 |
XM_006714416.3 | XP_006714479.1 | 5 - 6 | 1866 | 88 | |
XM_011532415.2 | XP_011530717.1 | 5 - 6 | 1778 | 88 | |
XM_011532416.2 | XP_011530718.1 | 5 - 6 | 1778 | 88 | |
XM_017008839.1 | XP_016864328.1 | 5 - 6 | 1778 | 88 | |
XM_017008840.1 | XP_016864329.1 | 5 - 6 | 1648 | 88 | |
XM_017008841.1 | XP_016864330.1 | 4 - 5 | 3489 | 88 | |
XM_017008842.1 | XP_016864331.1 | 4 - 5 | 1093 | 88 | |
XM_017008843.1 | XP_016864332.1 | 2 - 3 | 271 | 88 | |
XM_017008844.1 | XP_016864333.1 | 4 - 5 | 528 | 88 | |
GenBank mRNA | AF074924.1 | - | 5 - 6 | 1810 | 88 |
AF076605.1 | - | 4 - 5 | 1596 | 88 | |
AK295439.1 | - | 3 - 4 | 1418 | 88 | |
BC109309.1 | - | 5 - 6 | 1657 | 88 | |
BC109310.1 | - | 5 - 6 | 1657 | 88 | |
HQ234312.1 | - | 2 - 3 | 225 | 88 |