The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004784.2 | NP_004775.1 | 6 - 7 | 1948 | 157 |
XM_006714416.3 | XP_006714479.1 | 6 - 7 | 2004 | 157 | |
XM_017008839.1 | XP_016864328.1 | 6 - 7 | 1916 | 157 | |
XM_017008840.1 | XP_016864329.1 | 6 - 7 | 1786 | 157 | |
XM_017008841.1 | XP_016864330.1 | 5 - 6 | 3627 | 157 | |
XM_017008842.1 | XP_016864331.1 | 5 - 6 | 1231 | 157 | |
XM_017008843.1 | XP_016864332.1 | 3 - 4 | 409 | 157 | |
XM_017008844.1 | XP_016864333.1 | 5 - 6 | 666 | 157 | |
GenBank mRNA | AF074924.1 | - | 6 - 7 | 1948 | 157 |
AF076605.1 | - | 5 - 6 | 1734 | 157 | |
AK295439.1 | - | 4 - 5 | 1556 | 157 | |
BC109309.1 | - | 6 - 7 | 1795 | 157 | |
BC109310.1 | - | 6 - 7 | 1795 | 157 |