Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MEI1
- Entrez Gene ID
-
150365
- Gene Name
- meiotic double-stranded break formation protein 1
- Gene Aliases
-
SPATA38
- Chromosome Location
-
Chr.22: 41699503 - 41799456 on Build GRCh38
View Full Product Details