The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152513.3 | NP_689726.3 | 2 - 3 | 316 | 83 |
XM_011529935.1 | XP_011528237.1 | 2 - 3 | 325 | 83 | |
XM_011529936.1 | XP_011528238.1 | 2 - 3 | 325 | 83 | |
XM_011529937.1 | XP_011528239.1 | 2 - 3 | 325 | 83 | |
XM_011529938.1 | XP_011528240.1 | 2 - 3 | 325 | 83 | |
XM_011529939.1 | XP_011528241.1 | 2 - 3 | 325 | 83 | |
XM_011529940.1 | XP_011528242.1 | 2 - 3 | 325 | 83 | |
XM_011529941.1 | XP_011528243.1 | 2 - 3 | 325 | 83 | |
XM_011529942.2 | XP_011528244.1 | 2 - 3 | 516 | 83 | |
XM_011529943.1 | XP_011528245.1 | 2 - 3 | 325 | 83 | |
XM_011529944.1 | XP_011528246.1 | 2 - 3 | 325 | 83 | |
XM_011529945.2 | XP_011528247.1 | 2 - 3 | 327 | 83 | |
XM_011529946.1 | XP_011528248.1 | 2 - 3 | 325 | 83 | |
XM_011529947.1 | XP_011528249.1 | 2 - 3 | 325 | 83 | |
XM_011529950.1 | XP_011528252.1 | 2 - 3 | 325 | 83 | |
XM_011529955.1 | XP_011528257.1 | 2 - 3 | 325 | 83 | |
XM_011529956.2 | XP_011528258.1 | 2 - 3 | 325 | 83 | |
XM_017028633.1 | XP_016884122.1 | 2 - 3 | 325 | 83 | |
GenBank mRNA | BC025400.1 | - | 2 - 3 | 332 | 83 |
BC035720.1 | - | 2 - 3 | 315 | 83 |