Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LINC01166
- Entrez Gene ID
-
101927590
- Gene Name
- long intergenic non-protein coding RNA 1166
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 132943967 - 132965289 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_120627.1 |
|
1 - 2 |
103 |
75 |
GenBank mRNA |
AK095503.1 |
- |
1 - 2 |
103 |
75 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA483121.1 |
- |
1 - 2 |
102 |
|
Chr. 10 - 132945818 - 132965422 [-] on Build GRCh38 |
75 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 1167 |
LINC01167 |
TCONS_00018075 |
|
Sense |
Chr. - 132961340 - 132962237 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 1168 |
LINC01168 |
|
110.0
|
Anti-sense |
Chr. - 132965534 - 132976497 on Build GRCh38 |
View Full Product Details