Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH2D6
Entrez Gene ID
284948
Gene Name
SH2 domain containing 6
Gene Aliases
-
Chromosome Location
Chr.2: 85418931 - 85437029 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011532783.1 XP_011531085.1 4 - 5 599 58
XM_017003834.1 XP_016859323.1 7 - 8 4480 58
XM_017003835.1 XP_016859324.1 7 - 8 3907 58
XM_017003836.1 XP_016859325.1 6 - 7 4231 58
XM_017003837.1 XP_016859326.1 7 - 8 4349 58
XM_017003838.1 XP_016859327.1 6 - 7 3968 58
XM_017003839.1 XP_016859328.1 6 - 7 1523 58
XM_017003840.1 XP_016859329.1 6 - 7 4582 58
XM_017003841.1 XP_016859330.1 6 - 7 4601 58
XM_017003842.1 XP_016859331.1 8 - 9 4328 58
XM_017003843.1 XP_016859332.1 7 - 8 4213 58
XM_017003844.1 XP_016859333.1 8 - 9 3539 58
XM_017003845.1 XP_016859334.1 8 - 9 3981 58
XM_017003846.1 XP_016859335.1 8 - 9 4274 58
XM_017003847.1 XP_016859336.1 6 - 7 4583 58
XM_017003848.1 XP_016859337.1 6 - 7 4584 58
XM_017003849.1 XP_016859338.1 7 - 8 4259 58
XM_017003850.1 XP_016859339.1 7 - 8 4159 58
XM_017003853.1 XP_016859342.1 6 - 7 4601 58
XM_017003854.1 XP_016859343.1 6 - 7 4601 58
XM_017003855.1 XP_016859344.1 6 - 7 4601 58
XM_017003856.1 XP_016859345.1 6 - 7 4601 58
XM_017003857.1 XP_016859346.1 6 - 7 4601 58
XM_017003859.1 XP_016859348.1 4 - 5 320 58
GenBank mRNA BC110073.1 - 4 - 5 286 58

View Full Product Details