Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SH2D6
Entrez Gene ID
284948
Gene Name
SH2 domain containing 6
Gene Aliases
-
Chromosome Location
Chr.2: 85418931 - 85437029 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011532783.1 XP_011531085.1 5 - 6 622 60
XM_017003834.1 XP_016859323.1 8 - 9 4503 60
XM_017003835.1 XP_016859324.1 8 - 9 3930 60
XM_017003836.1 XP_016859325.1 7 - 8 4254 60
XM_017003837.1 XP_016859326.1 8 - 9 4372 60
XM_017003838.1 XP_016859327.1 7 - 8 3991 60
XM_017003839.1 XP_016859328.1 7 - 8 1546 60
XM_017003840.1 XP_016859329.1 7 - 8 4605 60
XM_017003841.1 XP_016859330.1 7 - 8 4624 60
XM_017003842.1 XP_016859331.1 9 - 10 4351 60
XM_017003843.1 XP_016859332.1 8 - 9 4236 60
XM_017003844.1 XP_016859333.1 9 - 10 3562 60
XM_017003845.1 XP_016859334.1 9 - 10 4004 60
XM_017003846.1 XP_016859335.1 9 - 10 4297 60
XM_017003847.1 XP_016859336.1 7 - 8 4606 60
XM_017003848.1 XP_016859337.1 7 - 8 4607 60
XM_017003849.1 XP_016859338.1 8 - 9 4282 60
XM_017003850.1 XP_016859339.1 8 - 9 4182 60
XM_017003853.1 XP_016859342.1 7 - 8 4624 60
XM_017003854.1 XP_016859343.1 7 - 8 4624 60
XM_017003855.1 XP_016859344.1 7 - 8 4624 60
XM_017003856.1 XP_016859345.1 7 - 8 4624 60
XM_017003857.1 XP_016859346.1 7 - 8 4624 60
XM_017003859.1 XP_016859348.1 5 - 6 343 60
GenBank mRNA BC110073.1 - 5 - 6 309 60

View Full Product Details