The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080464.2 | NP_001073933.2 | 9 - 10 | 1137 | 103 |
XM_005267590.4 | XP_005267647.1 | 9 - 10 | 1163 | 103 | |
XM_017021265.1 | XP_016876754.1 | 7 - 8 | 1604 | 103 | |
XM_017021266.1 | XP_016876755.1 | 7 - 8 | 1604 | 103 | |
XM_017021267.1 | XP_016876756.1 | 7 - 8 | 1604 | 103 | |
XM_017021268.1 | XP_016876757.1 | 9 - 10 | 1165 | 103 | |
XM_017021269.1 | XP_016876758.1 | 7 - 8 | 1604 | 103 | |
XM_017021270.1 | XP_016876759.1 | 7 - 8 | 1604 | 103 | |
XM_017021271.1 | XP_016876760.1 | 7 - 8 | 1604 | 103 | |
XM_017021272.1 | XP_016876761.1 | 7 - 8 | 1604 | 103 | |
XM_017021273.1 | XP_016876762.1 | 7 - 8 | 1604 | 103 | |
XM_017021274.1 | XP_016876763.1 | 9 - 10 | 1069 | 103 | |
XM_017021275.1 | XP_016876764.1 | 7 - 8 | 1604 | 103 | |
XM_017021276.1 | XP_016876765.1 | 7 - 8 | 1604 | 103 | |
GenBank mRNA | AX781433.1 | - | 9 - 10 | 1144 | 103 |
AX781435.1 | - | 9 - 10 | 1052 | 103 | |
BC035836.1 | - | 9 - 10 | 1112 | 103 | |
BC136637.1 | - | 9 - 10 | 1079 | 103 | |
BM544197.1 | - | 6 - 7 | 679 | 103 | |
BX247999.1 | - | 8 - 9 | 1886 | 103 |