The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017363.1 | NP_001017363.1 | 1 - 2 | 317 | 84 |
XM_017014283.1 | XP_016869772.1 | 3 - 4 | 3403 | 84 |