Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLCD2
Entrez Gene ID
727910
Gene Name
TLC domain containing 2
Gene Aliases
-
Chromosome Location
Chr.17: 1702790 - 1710368 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164407.1 NP_001157879.1 1 - 2 304 100

View Full Product Details