Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C7orf50
Entrez Gene ID
84310
Gene Name
chromosome 7 open reading frame 50
Gene Aliases
YCR016W
Chromosome Location
Chr.7: 978485 - 1138291 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_134538.1 1 - 2 170 95
GenBank mRNA BC025971.1 - 1 - 2 90 95

View Full Product Details