The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004231.3 | NP_004222.2 | 1 - 2 | 260 | 75 |
GenBank mRNA | AK312214.1 | - | 1 - 2 | 205 | 75 |
BC104230.1 | - | 1 - 2 | 211 | 75 | |
BC104231.1 | - | 1 - 2 | 211 | 75 | |
BC107854.1 | - | 1 - 2 | 237 | 75 | |
BG724326.1 | - | 1 - 2 | 245 | 75 | |
BU683155.1 | - | 1 - 2 | 75 | ||
CR456896.1 | - | 1 - 2 | 158 | 75 | |
D49400.1 | - | 1 - 2 | 208 | 75 | |
DB499437.1 | - | 1 - 2 | 260 | 75 |