The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271696.1 | NP_001258625.1 | 1 - 2 | 237 | 126 |
NM_001271697.1 | NP_001258626.1 | 1 - 2 | 237 | 126 | |
GenBank mRNA | AF038950.1 | - | 1 - 2 | 177 | 126 |
AF078777.1 | - | 1 - 2 | 157 | 126 | |
AF133659.1 | - | 1 - 2 | 169 | 126 | |
BX537833.1 | - | 1 - 2 | 176 | 126 | |
KU177874.1 | - | 1 - 2 | 169 | 126 | |
KU177875.1 | - | 1 - 2 | 169 | 126 |