The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271696.1 | NP_001258625.1 | 8 - 9 | 1092 | 148 |
NM_001271697.1 | NP_001258626.1 | 7 - 8 | 972 | 148 | |
NM_001271698.1 | NP_001258627.1 | 7 - 8 | 1014 | 148 | |
NM_001271699.1 | NP_001258628.1 | 7 - 8 | 975 | 148 | |
NM_004299.4 | NP_004290.2 | 8 - 9 | 1095 | 148 | |
GenBank mRNA | AF038950.1 | - | 8 - 9 | 1032 | 148 |
AF078777.1 | - | 8 - 9 | 1012 | 148 | |
AF133659.1 | - | 8 - 9 | 1024 | 148 | |
AK001418.1 | - | 8 - 9 | 1052 | 148 | |
AK294657.1 | - | 7 - 8 | 956 | 148 | |
AK307414.1 | - | 7 - 8 | 895 | 148 | |
BC006323.2 | - | 8 - 9 | 1028 | 148 | |
BT009918.1 | - | 8 - 9 | 1027 | 148 | |
BX537833.1 | - | 7 - 8 | 912 | 148 | |
KU177872.1 | - | 8 - 9 | 1027 | 148 | |
KU177873.1 | - | 8 - 9 | 1027 | 148 |