Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TLL2
Entrez Gene ID
7093
Gene Name
tolloid like 2
Gene Aliases
-
Chromosome Location
Chr.10: 96364606 - 96513926 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012465.3 NP_036597.1 3 - 4 605 57
GenBank mRNA AB023149.2 - 3 - 4 555 57
AF059516.1 - 3 - 4 728 57
BC013871.2 - 3 - 4 565 57
BC112341.1 - 3 - 4 488 57
BC112366.1 - 3 - 4 488 57
BC113577.1 - 3 - 4 421 57

View Full Product Details