Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TLL2
Entrez Gene ID
7093
Gene Name
tolloid like 2
Gene Aliases
-
Chromosome Location
Chr.10: 96364606 - 96513926 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012465.3 NP_036597.1 7 - 8 1163 97
GenBank mRNA AB023149.2 - 7 - 8 1113 97
AF059516.1 - 7 - 8 1286 97
BC013871.2 - 7 - 8 1123 97
BC112341.1 - 7 - 8 1046 97
BC112366.1 - 7 - 8 1046 97
BC113577.1 - 7 - 8 979 97

View Full Product Details