Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLIP3
Entrez Gene ID
25999
Gene Name
CAP-Gly domain containing linker protein 3
Gene Aliases
CLIPR-59, CLIPR59, RSNL1
Chromosome Location
Chr.19: 36014660 - 36032895 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199570.1 NP_001186499.1 8 - 9 1497 77
NM_015526.2 NP_056341.1 9 - 10 1413 77
GenBank mRNA AJ427922.1 - 9 - 10 1391 77
AK056445.1 - 7 - 8 896 77
AK090732.1 - 9 - 10 1416 77
AK092354.1 - 5 - 6 660 77
AK094738.1 - 9 - 10 1411 77
AK289509.1 - 8 - 9 1465 77
AK294748.1 - 9 - 10 1284 77
BC013116.1 - 9 - 10 1325 77
BC014486.1 - 6 - 7 856 77
BC128090.1 - 5 - 6 665 77

View Full Product Details