The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021959.2 | NP_068778.1 | 1 - 2 | 327 | 72 |
GenBank mRNA | AK313175.1 | - | 1 - 2 | 281 | 72 |
BC102010.1 | - | 1 - 2 | 266 | 72 | |
BC102011.1 | - | 1 - 2 | 266 | 72 | |
BC104750.1 | - | 1 - 2 | 266 | 72 | |
CB126242.1 | - | 1 - 2 | 394 | 72 | |
X81003.1 | - | 1 - 2 | 270 | 72 |