Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELP2
Entrez Gene ID
55250
Gene Name
elongator acetyltransferase complex subunit 2
Gene Aliases
SHINC-2, STATIP1, StIP
Chromosome Location
Chr.18: 36129874 - 36177931 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001324465.1 NP_001311394.1 6 - 7 652 144
NM_001324467.1 NP_001311396.1 6 - 7 652 144
NM_001324468.1 NP_001311397.1 5 - 6 573 144
NM_018255.3 NP_060725.1 6 - 7 652 144
NR_040110.2 6 - 7 652 144
NR_136898.1 6 - 7 652 144
NR_137173.1 6 - 7 652 144
GenBank mRNA AF403223.1 - 6 - 7 629 144
AK001741.1 - 6 - 7 602 144
AK222791.1 - 6 - 7 605 144
CR457269.1 - 6 - 7 592 144

View Full Product Details