The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015512.4 | NP_056327.4 | 47 - 48 | 7639 | 84 |
XM_017006129.1 | XP_016861618.1 | 49 - 50 | 10596 | 84 | |
XM_017006130.1 | XP_016861619.1 | 48 - 49 | 10526 | 84 | |
XM_017006131.1 | XP_016861620.1 | 49 - 50 | 10596 | 84 | |
XM_017006132.1 | XP_016861621.1 | 49 - 50 | 10597 | 84 | |
XM_017006133.1 | XP_016861622.1 | 49 - 50 | 10597 | 84 | |
GenBank mRNA | AB037831.3 | - | 47 - 48 | 7639 | 84 |
AB290163.1 | - | 47 - 48 | 7639 | 84 | |
AY221994.1 | - | 4 - 5 | 487 | 84 |