The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015512.4 | NP_056327.4 | 58 - 59 | 9626 | 83 |
XM_017006129.1 | XP_016861618.1 | 60 - 61 | 12583 | 83 | |
XM_017006130.1 | XP_016861619.1 | 59 - 60 | 12513 | 83 | |
XM_017006131.1 | XP_016861620.1 | 60 - 61 | 12583 | 83 | |
XM_017006132.1 | XP_016861621.1 | 60 - 61 | 12584 | 83 | |
XM_017006133.1 | XP_016861622.1 | 60 - 61 | 12584 | 83 | |
GenBank mRNA | AB037831.3 | - | 58 - 59 | 9626 | 83 |