The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001101421.3 | NP_001094891.3 | 14 - 15 | 1548 | 90 |
XM_011538223.2 | XP_011536525.1 | 14 - 15 | 1990 | 90 | |
XM_017019207.1 | XP_016874696.1 | 14 - 15 | 1990 | 90 | |
GenBank mRNA | DB339279.1 | - | 5 - 6 | 90 | |
DV080385.1 | - | 2 - 3 | 113 | 90 |