The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008895.2 | NP_001008895.1 | 18 - 19 | 2151 | 92 |
NM_001278513.1 | NP_001265442.1 | 18 - 19 | 2295 | 92 | |
NM_001278514.1 | NP_001265443.1 | 18 - 19 | 1963 | 92 | |
NM_003589.2 | NP_003580.1 | 18 - 19 | 1977 | 92 | |
XM_011537523.2 | XP_011535825.1 | 17 - 18 | 2511 | 92 | |
GenBank mRNA | AB012193.1 | - | 12 - 13 | 1325 | 92 |
AB178950.1 | - | 18 - 19 | 2028 | 92 | |
AF077188.1 | - | 18 - 19 | 1888 | 92 | |
AK296700.1 | - | 10 - 11 | 1272 | 92 | |
AK314835.1 | - | 18 - 19 | 2250 | 92 | |
AL833355.1 | - | 17 - 18 | 1915 | 92 | |
AY365124.1 | - | 18 - 19 | 2028 | 92 | |
BC008308.1 | - | 18 - 19 | 2039 | 92 |